Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007517.4 | NP_031543.2 | 3 | 555 | |
GenBank mRNA | AK144497.1 | - | 1 | 507 | |
AK154707.1 | - | 3 | 424 | ||
AK158035.1 | - | 3 | 346 | ||
AK167335.1 | - | 3 | 345 | ||
AK171011.1 | - | 3 | 358 | ||
AK171184.1 | - | 3 | 350 | ||
AK171202.1 | - | 3 | 364 | ||
AK171249.1 | - | 3 | 344 | ||
AK171541.1 | - | 3 | 345 | ||
AK171621.1 | - | 3 | 337 | ||
BC016485.1 | - | 3 | 348 | 3962982 | |
U41736.1 | - | 3 | 353 |