Gene Transcripts

Gene Symbol
Ackr3
Entrez Gene ID
12778
Gene Name
atypical chemokine receptor 3
Gene Aliases
AW541270, CXC-R7, CXCR-7, Cmkor1, Cxcr7, RDC-1, Rdc1
Chromosome Location
Chr. 1: 90203980 - 90215724 on Build GRCm38
UniGene ID
Mm.491219
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271607.1 NP_001258536.1 3 1989
NM_007722.4 NP_031748.2 2 1706
GenBank mRNA AF000236.1 - 2 1646
AK031100.1 - 1684
BC015254.1 - 2 1607 4242244

View Full Product Details