Gene Transcripts

Gene Symbol
Ctla2a
Entrez Gene ID
13024
Gene Name
cytotoxic T lymphocyte-associated protein 2 alpha
Gene Aliases
Ctla-2a
Chromosome Location
Chr. 13: 60934155 - 60936625 on Build GRCm38
UniGene ID
Mm.482089
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145799.1 NP_001139271.1 4, 5 462
NM_007796.2 NP_031822.2 4, 5 500
GenBank mRNA AK005471.1 - 4, 5 437
AK018801.1 - 4, 5 455
AK132290.1 - 4, 5 431
AK167433.1 - 4, 5 433
AK167664.1 - 4, 5 434
AY034577.1 - 4, 5 409
BC028437.1 - 4, 5 426 1244311
X15591.1 - 4, 5 409

Gene Symbol
Ctla2b
Entrez Gene ID
13025
Gene Name
cytotoxic T lymphocyte-associated protein 2 beta
Gene Aliases
AW545568, Ctla-2b
Chromosome Location
Chr. 13: 60895351 - 60897932 on Build GRCm38
UniGene ID
Mm.30144
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007797.1 NP_031823.1 4, 5 478
GenBank mRNA AY034578.1 - 4, 5 418
X15592.1 - 4, 5 418

View Full Product Details