Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009690.2 | NP_033820.2 | 6 | 1537 | |
GenBank mRNA | AF011428.1 | - | 6 | 1441 | |
AF018268.1 | - | 6 | 1476 | ||
AF018269.1 | - | 6 | 1456 | ||
AK159132.1 | - | 6 | 1486 | ||
AK159181.1 | - | 6 | 1540 | ||
AK159823.1 | - | 6 | 1505 | ||
BC006799.1 | - | 6 | 1478 | 3152586 | |
BC094459.1 | - | 6 | 1490 | 4161650 |