Gene Transcripts

Gene Symbol
Ctsd
Entrez Gene ID
13033
Gene Name
cathepsin D
Gene Aliases
CD, CatD
Chromosome Location
Chr. 7: 142375916 - 142387870 on Build GRCm38
UniGene ID
Mm.231395
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009983.2 NP_034113.1 7, 8 1034
GenBank mRNA AK089294.1 - 7, 8 992
AK150363.1 - 7, 8 995
AK150364.1 - 7, 8 992
AK150579.1 - 7, 8 992
AK150622.1 - 7, 8 992
AK150636.1 - 7, 8 992
AK150694.1 - 7, 8 992
AK150768.1 - 7, 8 992
AK150946.1 - 7, 8 1044
AK151256.1 - 7, 8 992
AK151275.1 - 7, 8 992
AK151276.1 - 7, 8 992
AK151374.1 - 7, 8 992
AK151829.1 - 7, 8 992
AK151962.1 - 7, 8 995
AK152036.1 - 7, 8 1005
AK152045.1 - 7, 8 1045
AK152168.1 - 7, 8 992
AK152187.1 - 7, 8 992
AK152188.1 - 7, 8 992
AK152406.1 - 7, 8 995
AK152458.1 - 7, 8 992
AK152567.1 - 7, 8 992
AK152578.1 - 7, 8 992
AK152593.1 - 7, 8 1022
AK152638.1 - 7, 8 1004
AK152680.1 - 7, 8 992
AK152828.1 - 7, 8 992
AK152881.1 - 7, 8 992
AK152892.1 - 7, 8 992
AK153191.1 - 7, 8 992
AK153291.1 - 7, 8 992
AK159097.1 - 7, 8 992
AK159209.1 - 7, 8 992
AK159440.1 - 7, 8 1045
AK159490.1 - 7, 8 992
AK159512.1 - 7, 8 1034
AK159580.1 - 7, 8 992
AK159642.1 - 5, 6 744
AK159743.1 - 7, 8 996
AK159896.1 - 7, 8 992
AK159970.1 - 7, 8 1045
AK165227.1 - 7, 8 1062
AK166864.1 - 7, 8 1005
AK166938.1 - 7, 8 1039
AK167143.1 - 7, 8 1030
AK167366.1 - 7, 8 1064
AK167698.1 - 7, 8 991
AK167750.1 - 7, 8 1005
AK167867.1 - 7, 8 993
AK169139.1 - 7, 8 1045
AK171195.1 - 7, 8 993
AK172583.1 - 7, 8 993
AK205399.1 - 2, 3 186
AK213803.1 - 2, 3 186
BC019682.1 - 3, 4 389
BC048900.1 - 3, 4 279
BC053937.1 - 3, 4 279
BC053940.1 - 3, 4 279
BC054758.1 - 7, 8 1064 6413315
BC057931.1 - 7, 8 1025 5253319
X52886.1 - 7, 8 1034
X53337.1 - 7, 8 974

View Full Product Details