Gene Transcripts

Gene Symbol
Anxa11
Entrez Gene ID
11744
Gene Name
annexin A11
Gene Aliases
A830099O17Rik, Anx11
Chromosome Location
Chr. 14: 25842151 - 25886804 on Build GRCm38
UniGene ID
Mm.294083
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013469.2 NP_038497.2 15 2255
GenBank mRNA AK137158.1 - 14 2565
AK140375.1 - 15 2229
BC012875.1 - 15 2175 3500318
U65986.1 - 15 2237

View Full Product Details