Gene Transcripts

Gene Symbol
Tmc1
Entrez Gene ID
13409
Gene Name
transmembrane channel-like gene family 1
Gene Aliases
4933416G09Rik, Beethoven, Bth, CWEA1, dn
Chromosome Location
Chr. 19: 20783458 - 21037152 on Build GRCm38
UniGene ID
Mm.219585
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028953.2 NP_083229.1 10, 11 1036
GenBank mRNA AF417579.1 - 10, 11 1036
AK033447.1 - 1, 2 33

View Full Product Details