Gene Transcripts

Gene Symbol
Slc25a1
Entrez Gene ID
13358
Gene Name
solute carrier family 25 (mitochondrial carrier, citrate transporter), member 1
Gene Aliases
1300019P08Rik, 2610100G11Rik, AI194714, Ctp, Dgsj, Slc20a3
Chromosome Location
Chr. 16: 17925211 - 17928219 on Build GRCm38
UniGene ID
Mm.229291
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153150.2 NP_694790.1 2 310
GenBank mRNA AK005070.1 - 2 240
AK159289.1 - 2 273
AK170191.1 - 2 274
BC037087.1 - 2 270 5124403

View Full Product Details