Gene Transcripts

Gene Symbol
Cuedc2
Entrez Gene ID
67116
Gene Name
CUE domain containing 2
Gene Aliases
3010002G01Rik
Chromosome Location
Chr. 19: 46329812 - 46338660 on Build GRCm38
UniGene ID
Mm.218848
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164290.1 NP_001157762.1 10 983
NM_001164291.1 NP_001157763.1 9 875
NM_001164292.1 NP_001157764.1 9 872
NM_001164293.1 NP_001157765.1 9 842
NM_001164294.1 NP_001157766.1 9 821
NM_001164295.1 NP_001157767.1 9 818
NM_024192.2 NP_077154.2 10 974
GenBank mRNA AB232359.1 - 8 773
AK003457.1 - 9 777
AK013884.1 - 9 833
BC003442.1 - 10 938 2650719

View Full Product Details