Gene Transcripts

Gene Symbol
Mcm10
Entrez Gene ID
70024
Gene Name
minichromosome maintenance deficient 10 (S. cerevisiae)
Gene Aliases
2410041F14Rik, AU018508, C330019M07Rik, C79164
Chromosome Location
Chr. 2: 4990724 - 5012848 on Build GRCm38
UniGene ID
Mm.23828
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027290.3 NP_081566.2 20 3438
GenBank mRNA AK010648.1 - 731
AK030780.1 - 20 3383
AK218102.1 - 1 341
BC023236.1 - 12 2300
BC039663.1 - 6 1288
BC064314.1 - 20 3801
BC094576.1 - 20 3438 6853824

View Full Product Details