Gene Transcripts

Gene Symbol
Cxxc5
Entrez Gene ID
67393
Gene Name
CXXC finger 5
Gene Aliases
4930415K17Rik
Chromosome Location
Chr. 18: 35828924 - 35861693 on Build GRCm38
UniGene ID
Mm.94560
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133687.2 NP_598448.1 3 1473
GenBank mRNA AK015150.1 - 3 1376
AK132950.1 - 3 1397
BC016207.1 - 2 1164 4912395
BC089314.1 - 3 1473 30541251

View Full Product Details