Gene Transcripts

Gene Symbol
Hopx
Entrez Gene ID
74318
Gene Name
HOP homeobox
Gene Aliases
1110018K11Rik, 1200015P04Rik, 2300002F06Rik, AI848177, AW490897, Cameo, Hdop, Hod, Hop, Ob1, Obl, Toto
Chromosome Location
Chr. 5: 77086986 - 77115123 on Build GRCm38
UniGene ID
Mm.181852
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159900.1 NP_001153372.1 3 927
NM_001159901.1 NP_001153373.1 2 1055
NM_175606.3 NP_783199.1 3 1087
GenBank mRNA AF492703.1 - 3 1084
AF492704.1 - 3 912
AK009007.1 - 3 844

View Full Product Details