Gene Transcripts

Gene Symbol
Olfr1367
Entrez Gene ID
258526
Gene Name
olfactory receptor 1367
Gene Aliases
MOR256-16, MOR256-65
Chromosome Location
Chr. 13: 21346930 - 21347880 on Build GRCm38
UniGene ID
Mm.377553
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146533.1 NP_666744.1 1 774
GenBank mRNA BC138110.1 - 1 794
BC138111.1 - 1 794 8861130

View Full Product Details