Gene Transcripts
- Gene Symbol
- Olfr275
- Entrez Gene ID
- 258857
- Gene Name
- olfactory receptor 275
- Gene Aliases
- MOR262-2
- Chromosome Location
- Chr. 4: 52825399 - 52826358 on Build GRCm38
- UniGene ID
- Mm.484340
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC101951.1 |
- |
1 |
595 |
|
BC141896.1 |
- |
1 |
588 |
|
View Full Product Details