Gene Transcripts

Gene Symbol
Olfr1346
Entrez Gene ID
258918
Gene Name
olfactory receptor 1346
Gene Aliases
MOR103-6
Chromosome Location
Chr. 7: 6474112 - 6475053 on Build GRCm38
UniGene ID
Mm.377748
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146916.1 NP_667127.1 1 286
GenBank mRNA BC137876.1 - 1 310
BC137877.1 - 1 310 8860896

View Full Product Details