Gene Transcripts

Gene Symbol
Tmem25
Entrez Gene ID
71687
Gene Name
transmembrane protein 25
Gene Aliases
0610039J01Rik, AI429491
Chromosome Location
Chr. 9: 44793772 - 44799273 on Build GRCm38
UniGene ID
Mm.41409
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027865.2 NP_082141.1 9 1907
GenBank mRNA AK002841.1 - 9 1908
AK090375.1 - 7 2181
AK154206.1 - 8 2393
AK165199.1 - 9 1939
AK165827.1 - 9 1892
BC060057.1 - 7 2230 5696102
BC060243.1 - 7 2230 5696102

View Full Product Details