Gene Transcripts

Gene Symbol
Tsga10
Entrez Gene ID
211484
Gene Name
testis specific 10
Gene Aliases
4933432N21Rik, AI843383, Gm217, Mtsga10
Chromosome Location
Chr. 1: 37728144 - 37866739 on Build GRCm38
UniGene ID
Mm.332756
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290720.1 NP_001277649.1 12 1619
NM_001290721.1 NP_001277650.1 11 1426
NM_207228.3 NP_997111.1 12 1486
GenBank mRNA AF530050.3 - 11 1431
AK030254.1 - 12 1619
AY618264.1 - 11 1431
BC053407.1 - 10 1254
BC066782.1 - 12 1483 6705127

View Full Product Details