Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011936.2 | NP_036066.2 | 3 | 639 | |
GenBank mRNA | AJ237917.1 | - | 3 | 625 | |
AK036677.1 | - | 3 | 629 | ||
AK040866.1 | - | 3 | 617 | ||
AK045465.1 | - | 3 | 616 | ||
AK046935.1 | - | 3 | 598 | ||
AK049502.1 | - | 3 | 632 | ||
AK088881.1 | - | 3 | 625 | ||
AK129437.1 | - | 3 | 606 | ||
AK161060.1 | - | 3 | 598 | ||
BC022222.1 | - | 3 | 590 | 4237261 | |
BC057008.1 | - | 3 | 605 | 5708558 |