Gene Transcripts

Gene Symbol
Sbk1
Entrez Gene ID
104175
Gene Name
SH3-binding kinase 1
Gene Aliases
Sbk
Chromosome Location
Chr. 7: 126272619 - 126294999 on Build GRCm38
UniGene ID
Mm.29660
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145587.2 NP_663562.1 2 636
GenBank mRNA BC024114.1 - 2 377
BC025837.1 - 2 473 5135723
BC031759.1 - 2 636 4194152

View Full Product Details