Gene Transcripts
- Gene Symbol
- RBBP4
- Entrez Gene ID
- 5928
- Gene Name
- RB binding protein 4, chromatin remodeling factor
- Gene Aliases
- NURF55, RBAP48, lin-53
- Chromosome Location
- Chr. 1: 32651148 - 32686211 on Build GRCh38
- UniGene ID
- Hs.16003
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135255.1 |
NP_001128727.1
|
8, 9 |
1115 |
|
NM_001135256.1 |
NP_001128728.1
|
8, 9 |
1018 |
|
NM_005610.2 |
NP_005601.1
|
8, 9 |
1118 |
|
GenBank mRNA |
AK222779.1 |
- |
8, 9 |
1021 |
|
AK299251.1 |
- |
8, 9 |
1018 |
|
AK299410.1 |
- |
6, 7 |
729 |
|
AK312571.1 |
- |
8, 9 |
1056 |
|
BC003092.1 |
- |
8, 9 |
1045 |
3356558 |
BC015123.1 |
- |
6, 7 |
779 |
|
BC053904.1 |
- |
8, 9 |
1031 |
6526200 |
BC075836.1 |
- |
8, 9 |
986 |
6338602 |
BT007309.1 |
- |
8, 9 |
959 |
|
X71810.1 |
- |
8, 9 |
1032 |
|
X74262.1 |
- |
8, 9 |
1043 |
|
View Full Product Details