Gene Transcripts
- Gene Symbol
- Myo3b
- Entrez Gene ID
- 329421
- Gene Name
- myosin IIIB
- Gene Aliases
- A430065P19Rik
- Chromosome Location
- Chr. 2: 70039029 - 70429195 on Build GRCm38
- UniGene ID
- Mm.99648
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_177376.4 |
NP_796350.2
|
35 |
4912 |
|
GenBank mRNA |
AK040119.1 |
- |
10 |
2065 |
|
View Full Product Details