Gene Transcripts

Gene Symbol
GPA33
Entrez Gene ID
10223
Gene Name
glycoprotein A33
Gene Aliases
A33
Chromosome Location
Chr. 1: 167052836 - 167090631 on Build GRCh38
UniGene ID
Hs.651244
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005814.2 NP_005805.1 2 511
XM_017000005.1 XP_016855494.1 3 339
GenBank mRNA AK312833.1 - 2 474
BC069705.1 - 2 511 7262529
BC069723.1 - 2 511 7262541
BC069745.1 - 2 511 7262553
BC069761.1 - 2 511 7262565
BC069789.1 - 2 511 7262482
BC074830.2 - 2 189 30915543
BC074876.2 - 2 189 30915236
BC107164.1 - 2 466 40034324
BC107165.1 - 2 468 40034331
U79725.1 - 2 511

View Full Product Details