Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177910.3 | NP_808578.1 | 9 | 1301 | |
GenBank mRNA | AK088295.1 | - | 9 | 1283 | |
AK148125.1 | - | 1301 | |||
AK158470.1 | - | 9 | 1301 | ||
AK165816.1 | - | 8 | 1386 | ||
BC061207.1 | - | 10 | 1175 | 30249330 |