Gene Transcripts
- Gene Symbol
- Fastkd5
- Entrez Gene ID
- 380601
- Gene Name
- FAST kinase domains 5
- Gene Aliases
- C78212, mKIAA1792
- Chromosome Location
- Chr. 2: 130613838 - 130630027 on Build GRCm38
- UniGene ID
- Mm.27090
- Species
- Mus musculus
- Gene Symbol
- Ubox5
- Entrez Gene ID
- 140629
- Gene Name
- U box domain containing 5
- Gene Aliases
- 1500010O06Rik, C330018L13Rik, Rnf37, UIP5, Ubce7ip5, Ube7ip5
- Chromosome Location
- Chr. 2: 130589996 - 130630038 on Build GRCm38
- UniGene ID
- Mm.486003
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK170024.1 |
- |
2 |
1035 |
|
View Full Product Details