Gene Transcripts

Gene Symbol
Map6d1
Entrez Gene ID
208158
Gene Name
MAP6 domain containing 1
Gene Aliases
A330066I15Rik
Chromosome Location
Chr. 16: 20233309 - 20241358 on Build GRCm38
UniGene ID
Mm.326383
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198599.2 NP_941001.2 2 447
GenBank mRNA AK079603.1 - 2 410
AK163656.1 - 2 411
BB653005.1 - 2 448
BC116220.1 - 2 410 40091320
BC116221.1 - 2 410 40091321
BE979311.1 - 2 121

View Full Product Details