Gene Transcripts

Gene Symbol
Ttc7
Entrez Gene ID
225049
Gene Name
tetratricopeptide repeat domain 7
Gene Aliases
1110035E02Rik, 1700007L07Rik, Ttc7a, fsn, hea
Chromosome Location
Chr. 17: 87282082 - 87381770 on Build GRCm38
UniGene ID
Mm.77396
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028639.3 NP_082915.1 19 2653
GenBank mRNA AK040578.1 - 19 2656
AK040610.1 - 19 2624
AK050263.1 - 19 2640
AK080284.1 - 19 2632
AK153704.1 - 19 2635
AK165244.1 - 12 1315
BC023773.1 - 15 1611
BC025613.1 - 1 136
BC042512.1 - 19 2330 4223801
BC049254.1 - 3 376
BC057906.1 - 2 276

View Full Product Details