Gene Transcripts

Gene Symbol
Cecr5
Entrez Gene ID
214932
Gene Name
cat eye syndrome chromosome region, candidate 5
Gene Aliases
A930002G03Rik
Chromosome Location
Chr. 6: 120509494 - 120531364 on Build GRCm38
UniGene ID
Mm.255322
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144815.2 NP_659064.1 7 794
GenBank mRNA AK140387.1 - 7 815
BC014705.1 - 7 803 4238201

View Full Product Details