Gene Transcripts

Gene Symbol
MAP2K3
Entrez Gene ID
5606
Gene Name
mitogen-activated protein kinase kinase 3
Gene Aliases
MAPKK3, MEK3, MKK3, PRKMK3, SAPKK-2, SAPKK2
Chromosome Location
Chr. 17: 21284656 - 21315240 on Build GRCh38
UniGene ID
Hs.514012
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316332.1 NP_001303261.1 10 1127
NM_002756.4 NP_002747.2 9 744
NM_145109.2 NP_659731.1 9 1002
XM_005256723.2 XP_005256780.1 10 1042
XM_011523958.2 XP_011522260.1 9 801
XM_011523959.1 XP_011522261.1 11 1009
XM_017024857.1 XP_016880346.1 9 871
XM_017024858.1 XP_016880347.1 10 859
XM_017024859.1 XP_016880348.1 11 1199
GenBank mRNA AB451226.1 - 9 737
AB451349.1 - 9 737
AK093838.1 - 10 1046
AK223490.1 - 9 877
AK315505.1 - 9 947
BC032478.1 - 9 937 5215093
CR536514.1 - 9 737
CR541793.1 - 9 737
D87116.1 - 9 794
GQ225578.1 - 9 737
L36719.1 - 11 987
U66839.1 - 9 863

View Full Product Details