Gene Transcripts
- Gene Symbol
- Spidr
- Entrez Gene ID
- 224008
- Gene Name
- scaffolding protein involved in DNA repair
- Gene Aliases
- 2310008H04Rik, Kiaa0146
- Chromosome Location
- Chr. 16: 15889224 - 16146851 on Build GRCm38
- UniGene ID
- Mm.244703
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_146068.4 |
NP_666180.2
|
14, 15 |
2108 |
|
GenBank mRNA |
AK044836.1 |
- |
14, 15 |
2091 |
|
AK052832.1 |
- |
14, 15 |
2071 |
|
AK075871.1 |
- |
14, 15 |
2073 |
|
AK166448.1 |
- |
9, 10 |
1403 |
|
AK170784.1 |
- |
10, 11 |
1740 |
|
AK172902.1 |
- |
14, 15 |
2061 |
|
BC026877.1 |
- |
4, 5 |
442 |
|
BC046558.1 |
- |
8, 9 |
1112 |
|
BC069859.1 |
- |
5, 6 |
661 |
|
BC085258.1 |
- |
5, 6 |
479 |
|
BC089026.1 |
- |
14, 15 |
2045 |
30615298 |
View Full Product Details