Gene Transcripts

Gene Symbol
Usp1
Entrez Gene ID
230484
Gene Name
ubiquitin specific peptidase 1
Gene Aliases
Chromosome Location
Chr. 4: 98923622 - 98935543 on Build GRCm38
UniGene ID
Mm.371692
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301414.1 NP_001288343.1 9 2428
NM_146144.4 NP_666256.2 9 2409
GenBank mRNA AK080882.1 - 9 2410
BC018179.1 - 9 2289 3585191
BC020007.1 - 9 2261 3980469
BC023812.1 - 6 1583

View Full Product Details