Gene Transcripts

Gene Symbol
Wipf1
Entrez Gene ID
215280
Gene Name
WAS/WASL interacting protein family, member 1
Gene Aliases
AI115543, D2Ertd120e, WIP, Waspip
Chromosome Location
Chr. 2: 73429610 - 73529733 on Build GRCm38
UniGene ID
Mm.223504
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289722.1 NP_001276651.1 8 2433
NM_001289723.1 NP_001276652.1 8 2398
NM_153138.4 NP_694778.1 8 2428
GenBank mRNA AK031046.1 - 8 2395
AK156547.1 - 8 2433
AK168092.1 - 4 1582
BC049788.1 - 8 2351 6514406

View Full Product Details