Gene Transcripts

Gene Symbol
Ttc24
Entrez Gene ID
214191
Gene Name
tetratricopeptide repeat domain 24
Gene Aliases
A430025D11Rik
Chromosome Location
Chr. 3: 88069410 - 88081987 on Build GRCm38
UniGene ID
Mm.229191
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172526.3 NP_766114.1 5 1144
GenBank mRNA AK039884.1 - 5 1145
BC147554.1 - 4 848 9056545
BC147555.1 - 4 848 9056546

View Full Product Details