Gene Transcripts
- Gene Symbol
- Ttc24
- Entrez Gene ID
- 214191
- Gene Name
- tetratricopeptide repeat domain 24
- Gene Aliases
- A430025D11Rik
- Chromosome Location
- Chr. 3: 88069410 - 88081987 on Build GRCm38
- UniGene ID
- Mm.229191
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_172526.3 |
NP_766114.1
|
10 |
2070 |
|
GenBank mRNA |
AK039884.1 |
- |
10 |
2071 |
|
View Full Product Details