Gene Transcripts

Gene Symbol
Ybey
Entrez Gene ID
216119
Gene Name
ybeY metallopeptidase
Gene Aliases
1110068N23Rik, A130042E20Rik, C21orf57, ORF57, ORF66
Chromosome Location
Chr. 10: 76459567 - 76469114 on Build GRCm38
UniGene ID
Mm.344778
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172550.4 NP_766138.3 5 1068
GenBank mRNA AK037727.1 - 5 795
AK136543.1 - 5 819
BC057308.1 - 5 1068 5703033

View Full Product Details