Gene Transcripts

Gene Symbol
Wdr41
Entrez Gene ID
218460
Gene Name
WD repeat domain 41
Gene Aliases
B830029I03Rik, MSTP048
Chromosome Location
Chr. 13: 94976213 - 95023316 on Build GRCm38
UniGene ID
Mm.33856
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK082348.1 - 10 2074
BC138095.1 - 10 1563 8861114
BC145143.1 - 10 1613
BC145144.1 - 10 1563 9053695

View Full Product Details