Gene Transcripts

Gene Symbol
Rsbn1
Entrez Gene ID
229675
Gene Name
rosbin, round spermatid basic protein 1
Gene Aliases
C230004D03Rik, E330004N01, Rosbin, Rsbn, Rsbp, mKIAA3002
Chromosome Location
Chr. 3: 103914067 - 103966620 on Build GRCm38
UniGene ID
Mm.29366
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172684.2 NP_766272.2 2 1287
GenBank mRNA AB101658.1 - 2 1289
AK087685.1 - 1 253
AK122577.1 - 2 822
AK134093.1 - 2 287
AK138603.1 - 2 831
BC094667.1 - 2 812
BC139213.1 - 2 1256 8862235
BC139215.1 - 2 1256 8862237

View Full Product Details