Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172692.3 | NP_766280.2 | 3, 4 | 1062 | |
GenBank mRNA | AK089192.1 | - | 3, 4 | 748 | |
AK158174.1 | - | 3, 4 | 1062 | ||
AK173213.1 | - | 3, 4 | 1048 | ||
BC031397.1 | - | 3, 4 | 735 | ||
BC034105.1 | - | 3, 4 | 287 | ||
BC056935.1 | - | 3, 4 | 585 | 5703478 |