Gene Transcripts
- Gene Symbol
- Synrg
- Entrez Gene ID
- 217030
- Gene Name
- synergin, gamma
- Gene Aliases
- Ap1gbp1, SYNG
- Chromosome Location
- Chr. 11: 83964380 - 84044578 on Build GRCm38
- UniGene ID
- Mm.441722
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_194341.2 |
NP_919322.1
|
11 |
1329 |
|
GenBank mRNA |
BC056370.1 |
- |
11 |
1289 |
|
View Full Product Details