Gene Transcripts

Gene Symbol
D17H6S56E-5
Entrez Gene ID
110956
Gene Name
DNA segment, Chr 17, human D6S56E 5
Gene Aliases
G7e
Chromosome Location
Chr. 17: 34996678 - 35000741 on Build GRCm38
UniGene ID
Mm.22506
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK037701.1 - 2 760
AK075966.1 - 2 783
AK161228.1 - 2 758
BC027314.1 - 2 746 3985942
L78788.1 - 2 1151

View Full Product Details