Gene Transcripts

Gene Symbol
Foxp1
Entrez Gene ID
108655
Gene Name
forkhead box P1
Gene Aliases
3110052D19Rik, 4932443N09Rik, AI461938, AW494214
Chromosome Location
Chr. 6: 98925342 - 99523024 on Build GRCm38
UniGene ID
Mm.234965
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001197321.1 NP_001184250.1 3 335
NM_053202.2 NP_444432.1 4 391
GenBank mRNA AA451299.1 - 3 334
AB221601.1 - 1 80
AB221602.1 - 1 80
AB594192.1 - 5 796
AF339103.1 - 1 80
AF339104.1 - 1 80
AK040800.1 - 4 467
AK171994.1 - 3 332
BC064764.1 - 3 321 30430449
BC065386.1 - 3 335

View Full Product Details