Gene Transcripts

Gene Symbol
ST8SIA2
Entrez Gene ID
8128
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 2
Gene Aliases
HsT19690, SIAT8B, ST8SIA-II, STX
Chromosome Location
Chr. 15: 92393910 - 92468728 on Build GRCh38
UniGene ID
Hs.302341
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006011.3 NP_006002.1 2 278
XM_017022642.1 XP_016878131.1 2 441
GenBank mRNA AK313924.1 - 2 226
AW163487.1 - 278
BC069584.1 - 2 147
BC096202.3 - 2 147 40004644
BC096203.3 - 2 147 40004645
BC096204.3 - 2 147 40004646
U33551.1 - 2 123
U82762.1 - 2 148

View Full Product Details