Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001166456.1 | NP_001159928.1 | 13 | 1355 | |
NM_001166458.1 | NP_001159930.1 | 13 | 1562 | ||
NM_134086.4 | NP_598847.2 | 13 | 2048 | ||
GenBank mRNA | AF184240.2 | - | 12 | 1140 | |
AK034130.1 | - | 13 | 2050 | ||
AK035098.1 | - | 12 | 1067 | ||
AK047459.1 | - | 13 | 1579 | ||
AK049294.1 | - | 13 | 1328 | ||
AK081724.1 | - | 13 | 1564 | ||
AK082683.1 | - | 13 | 1266 | ||
AK162612.1 | - | 13 | 1413 | ||
AK163914.1 | - | 8 | 666 | ||
AK165188.1 | - | 13 | 1615 | ||
BC030378.1 | - | 13 | 1264 | 5375319 |