Gene Transcripts

Gene Symbol
Slc38a1
Entrez Gene ID
105727
Gene Name
solute carrier family 38, member 1
Gene Aliases
AA408026, AA409865, AL022800, AU015942, NAT2
Chromosome Location
Chr. 15: 96571418 - 96642913 on Build GRCm38
UniGene ID
Mm.103568
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166456.1 NP_001159928.1 7 829
NM_001166458.1 NP_001159930.1 7 1036
NM_134086.4 NP_598847.2 7 1522
GenBank mRNA AF184240.2 - 6 614
AK029189.1 - 7 699
AK034130.1 - 7 1524
AK035098.1 - 6 541
AK047459.1 - 7 1053
AK049173.1 - 7 736
AK049294.1 - 7 802
AK050914.1 - 7 737
AK081724.1 - 7 1038
AK162612.1 - 7 887
AK163914.1 - 2 140
AK165188.1 - 7 1089
AK197673.1 - 2 135
BC030378.1 - 7 738 5375319

View Full Product Details