Gene Transcripts

Gene Symbol
Timd2
Entrez Gene ID
171284
Gene Name
T cell immunoglobulin and mucin domain containing 2
Gene Aliases
AA816106, AU019358, AU019662, C78111, G9D3, TIM-2, Tim2
Chromosome Location
Chr. 11: 46668960 - 46707104 on Build GRCm38
UniGene ID
Mm.234654
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161355.1 NP_001154827.1 3 431
NM_001161356.1 NP_001154828.1 3 204
NM_134249.5 NP_599010.4 3 313
GenBank mRNA AB009015.1 - 3 204
AF399827.1 - 3 165
AF399828.1 - 3 165
AK145630.1 - 3 192
AK163225.1 - 3 192
AK166877.1 - 3 185
BC025096.1 - 3 243 4158605
BC028829.1 - 3 275 5102129
BY731803.1 - 3 313
CB953226.1 - 3 434

View Full Product Details