Gene Transcripts

Gene Symbol
Abcg4
Entrez Gene ID
192663
Gene Name
ATP-binding cassette, sub-family G (WHITE), member 4
Gene Aliases
6430517O04Rik
Chromosome Location
Chr. 9: 44273188 - 44289102 on Build GRCm38
UniGene ID
Mm.101876
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138955.3 NP_620405.3 15 3132
GenBank mRNA AF411084.1 - 15 3009
AF425077.1 - 15 3010
AJ426047.2 - 15 3010
AK018283.1 - 1 932
AK043147.1 - 15 3133
AK157317.1 - 15 3457
BC016200.1 - 15 3012 4511456
BC023077.1 - 9 2220
BC030301.1 - 1 1328
BC059033.1 - 6 2304

View Full Product Details