Gene Transcripts

Gene Symbol
Pamr1
Entrez Gene ID
210622
Gene Name
peptidase domain containing associated with muscle regeneration 1
Gene Aliases
5930437L24, E430002G05Rik, Ramp
Chromosome Location
Chr. 2: 102550021 - 102643041 on Build GRCm38
UniGene ID
Mm.28649
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173749.4 NP_776110.3 9 1287
GenBank mRNA AK088017.1 - 9 1186
BC031841.1 - 9 1241 3675412
BC057685.1 - 9 1241 3675412

View Full Product Details