Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_175431.4 | NP_780640.2 | 2 | 479 | |
GenBank mRNA | AK044133.1 | - | 2 | 480 | |
AK050447.1 | - | 2 | 445 | ||
BC100429.1 | - | 2 | 258 | 30917787 | |
BC144941.1 | - | 2 | 210 | ||
BC145648.1 | - | 2 | 210 | 40130951 |