Gene Transcripts

Gene Symbol
Rrp8
Entrez Gene ID
101867
Gene Name
ribosomal RNA processing 8, methyltransferase, homolog (yeast)
Gene Aliases
1500003O22Rik, 2900001K19Rik, AW538116
Chromosome Location
Chr. 7: 105731730 - 105737385 on Build GRCm38
UniGene ID
Mm.231400
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025897.2 NP_080173.1 7 1745
NM_133951.1 NP_598712.1 7 1779
GenBank mRNA AK005138.1 - 7 1661
AK013462.1 - 7 1655
AK045361.1 - 7 1597
AK077786.1 - 7 1668
AK154337.1 - 7 1772
AK154858.1 - 7 1720
AK165828.1 - 7 1593
AK172189.1 - 7 1618
AK172224.1 - 7 1618
AK200577.1 - 1 259
BC022923.1 - 7 1677 4242458
BC046799.1 - 7 1652 6403599

View Full Product Details