Gene Transcripts

Gene Symbol
Arfip2
Entrez Gene ID
76932
Gene Name
ADP-ribosylation factor interacting protein 2
Gene Aliases
2310002N04Rik, POR, Por1
Chromosome Location
Chr. 7: 105634201 - 105640416 on Build GRCm38
UniGene ID
Mm.41637
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029802.4 NP_084078.3 3 349
GenBank mRNA AK009100.1 - 3 287
AK154772.1 - 3 315
BC013794.1 - 3 260 2648132
BC022942.1 - 3 273 3583149
BC028310.1 - 3 247
BC034520.1 - 3 330 3964827
BC079603.1 - 1 845

View Full Product Details