Gene Transcripts

Gene Symbol
Rsph6a
Entrez Gene ID
83434
Gene Name
radial spoke head 6 homolog A (Chlamydomonas)
Gene Aliases
RSP4, Rshl1
Chromosome Location
Chr. 7: 19054678 - 19074447 on Build GRCm38
UniGene ID
Mm.84435
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031255.2 NP_112545.2 3 1002
GenBank mRNA AF329192.1 - 3 995
AK029699.1 - 3 1002
BC137992.1 - 3 995 8861011

View Full Product Details